missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Babcock, specific gravity 1.825, Ricca Chemical

APHA for Fat, Babcock Method

Supplier:  Ricca Chemical Company 819232

 View more versions of this product

Catalog No. 819232


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
specific gravity 1.825
94.0,4.31
Safety-coated Glass Bottle
H2O4S
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Laboratory
Colorless
7664-93-9,7732-18-5
98.0,4.49
1 L
Liquid
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions