missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Babcock, specific gravity 1.825, Ricca Chemical
APHA for Fat, Babcock Method
Supplier: Ricca Chemical Company 81921
Specifications
| Sulfuric Acid | |
| specific gravity 1.825 | |
| 94.0,4.31 | |
| Safety-coated Amber Glass Bottle | |
| H2O4S | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| Laboratory |
| Colorless | |
| 7664-93-9,7732-18-5 | |
| 98.0,4.49 | |
| 4 L | |
| Liquid | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |