missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, for analysis, ca. 96% solution in water

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

$101.51 - $307.95

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124645000
SDS View Documents
Thermo Scientific Chemicals
124645000
500 mL Glass Bottle
Each for $101.51
Only null left
 
AC124645001
SDS View Documents
Thermo Scientific Chemicals
124645001
500 mL Plastic bottle
Each for $107.74
Only null left
 
AC124640011
SDS View Documents
Thermo Scientific Chemicals
124640011
1 L Plastic bottle
Each for $161.79
Only null left
 
AC124640025
SDS View Documents
Thermo Scientific Chemicals
124640025
2.5 L Glass Bottle
Each for $307.95
Case of 4 Each for $0.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O

Specifications

7664-93-9
Colorless
290.0°C
Glass Bottle
H2SO4
500 mL
1.83
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
CHEBI:26836
95 to 97%
Liquid
10.0°C
1.8300g/mL
0.0005% max.
H2O4S
MFCD00064589
01,470; 05,633; 06,558; 07,347; 09,441
15,9104
Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
OS(O)(=O)=O
98.07
21 mPa.s (25°C)
98.08
Analysis
Sulfuric acid
Videos
Safety and Handling

Safety and Handling

Danger

SDS
Documents

Documents

RUO – Research Use Only