missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, ACS Reagent, Reagents
Sulfuric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C5102640500A
Specifications
| 0 | |
| Colorless | |
| 330°C | |
| 1.84 | |
| H2O4S | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| ACS |
| Sulfuric Acid, ACS Reagent | |
| 7664-93-9,7732-18-5 | |
| Poly Bottle | |
| 500 mL | |
| Liquid | |
| Oil of Vitriol, Sulphuric Acid, Dihydrogen Sulfate | |
| OS(O)(=O)=O | |
| 98.07 |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| Oil of Vitriol, Sulphuric Acid, Dihydrogen Sulfate | |
| OS(O)(=O)=O |