missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, ACS Reagent, 95.0 to 98.0%, Honeywell Fluka™

Catalog No. 6001586
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass Bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001586 2.5 L Plastic Bottle
6001582 100 mL Glass Bottle
6001590 500 mL Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001586 Supplier Honeywell Inc Supplier No. 2581052.5L
Only null left

ACS Reagent, 95.0-98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Density 1.840g/mL at 25°C
Packaging Plastic Bottle
Residue after Ignition ≤5ppm
Vapor Pressure 1mmHg (146°C)
Quantity 2.5 L
Linear Formula H2SO4
UN Number 1830
Beilstein 2037554
Formula Weight 98.08g/mol
Percent Purity 95 to 98%
Grade ACS Reagent
Chemical Name or Material Sulfuric Acid
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H290-H314
EINECSNumber 231-639-5
RTECSNumber WS5600000