missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid ACS, 95 to 98%, MilliporeSigma™
Supplier: Technidata SX12445
Specifications
| Sulfuric Acid | |
| -20°C | |
| 335°C | |
| Glass Bottle | |
| 2.5 L | |
| H2SO4 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| 95 to 98% |
| Suprapur™ | |
| 7664-93-9 | |
| 1.84 g/cm3 (20°C) | |
| 0.0001 hPa (20°C) | |
| H2O4S | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.08 | |
| ACS |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Safety and Handling
EINECSNumber : 231-639-5
RTECSNumber : WS5600000
Recommended Storage : Store without restrictions