missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid ACS, 95 to 98%, MilliporeSigma™

Catalog No. MSX1244PC5
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Poly Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MSX1244PC5 500 mL Poly Coated Glass Bottle
MSX12446 500 mL Glass Bottle
MSX12445 2.5 L Glass Bottle
23749903 2.5 L Poly Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MSX1244PC5 Supplier Technidata Supplier No. SX1244PC5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Melting Point -20°C
Boiling Point 335°C
Density 1.84 g/cm3 (20°C)
Packaging Poly Coated Glass Bottle
Vapor Pressure 0.0001 hPa (20°C)
Quantity 500 mL
Linear Formula H2SO4
Formula Weight 98.08
Percent Purity 95 to 98%
Grade ACS
Chemical Name or Material Sulfuric Acid, Suprapur™
Show More Show Less
EINECSNumber 231-639-5
RTECSNumber WS5600000
Recommended Storage Store without restrictions