missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid ACS, 95 to 98%, MilliporeSigma™

Catalog No. 23749903
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Poly Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
23749903 2.5 L Poly Coated Glass Bottle
MSX12446 500 mL Glass Bottle
MSX1244PC5 500 mL Poly Coated Glass Bottle
MSX12445 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23749903 Supplier Technidata Supplier No. SX124475

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Melting Point -20°C
Boiling Point 335°C
Density 1.84 g/cm3 (20°C)
Packaging Poly Coated Glass Bottle
Vapor Pressure 0.0001 hPa (20°C)
Quantity 2.5 L
Linear Formula H2SO4
Formula Weight 98.08
Percent Purity 95 to 98%
Grade ACS
Chemical Name or Material Sulfuric Acid, Suprapur™
Show More Show Less
EINECSNumber 231-639-5
RTECSNumber WS5600000
Recommended Storage Store without restrictions