Learn More
Sulfuric acid, ACS, 95.0-98.0%
Description
Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.84 |
| Assay Percent Range | 95.0 to 98.0% |
| Quantity | 4 x 1 kg |
| Physical Form | Liquid |
| Solubility Information | Sulfuric acid is miscible with water at all concentrations. |
| Formula Weight | 98.08 |
| Grade | ACS |
| Chemical Name or Material | Sulfuric Acid |
Safety and Handling
| Hazard Category | H314-H330-H350 |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
| Precautionary Statement | P201-P202-P260-P264b-P271-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P363-P501c |
| DOTInformation | DOT Class: 8; Packaging Group: II |
| EINECSNumber | 231-639-5 |
| RTECSNumber | WS5600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only