missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, ACS, 95.0-98.0%

Catalog No. AA33273A1
Change view
Click to view available options
Quantity:
1 kg
4 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33273A1 1 kg
AA33273D6 4 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33273A1 Supplier Thermo Scientific Chemicals Supplier No. 33273A1

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Density 1.84
Assay Percent Range 95.0 to 98.0%
Quantity 1 kg
Physical Form Liquid
Solubility Information Sulfuric acid is miscible with water at all concentrations.
Formula Weight 98.08
Grade ACS
Chemical Name or Material Sulfuric Acid
Hazard Category H314-H330-H350
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P201-P202-P260-P264b-P271-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: II
EINECSNumber 231-639-5
RTECSNumber WS5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only