missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid 96% LM Prime, Transene

Catalog No. p-200106338
Change view
Click to view available options
Quantity:
55 Gallon
2.5 L
1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502744718 2.5 L
502744723 55 Gallon
502744724 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 502744718 Supplier Doe and Ingalls Management LLC Supplier No. T07019212.5L
Only null left

Sulfuric acid is used in semiconductor processing to remove residual organic contamination prior to metalization steps

Designed for electronics manufacturing are referred to as Electronic Grade solvents. These solvents contain very low levels of heavy metals, typically measured in parts per billion or parts per trillion. They are commonly utilized as cleaners and etchants in the production of semiconductors and microprocessors. 96% Sulfuric Acid Electronic/Cleanroom Grade is suitable for use in electronics and related fields.

  • Industries:
  • Electronics and Semiconductor
  • Cleaning agents

Safety and Shipping Information: Sulfuric Acid from Lab Alley is classified as hazardous, and a hazmat fee is required. DOT: Sulfuric Acid, 8, UN1830, PG II

Chemical Identifiers

CAS 7664-93-9
Molecular Weight (g/mol) 98 g/mol

Specifications

Packaging HDPE
Quantity 2.5 L
Percent Purity 96%
Grade Electronic
Chemical Name or Material Sulfuric Acid