missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 90% (w/w), Ricca Chemical
Supplier: Ricca Chemical Company R81940004A
Specifications
| Sulfuric Acid | |
| 7664-93-9,7732-18-5 | |
| 95.0,6.0 | |
| 4 L | |
| Liquid | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Laboratory |
| Colorless | |
| 94.0,5.0 | |
| Natural Poly Bottle | |
| H2O4S | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 90% W/W | |
| 90% (w/w) Aqueous Solution |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Safety and Handling
ShelfLife : 24 Months