missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 66 °Be, Technical, Approx. 93%, Spectrum™ Chemical

Catalog No. p-7158340
Change view
Click to view available options
Quantity:
2.5 L
20 L
200 L
Packaging:
Drum
Glass Bottle
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607155 2.5 L Glass Bottle
18607152 20 L Poly Drum
18607154 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607155 Supplier Spectrum Chemical Mfg Cor Supplier No. S17352.5LTGL
Only null left

S1735, 7664-93-9, H2SO4

Spectrum™ Chemical Sulfuric Acid, 66 °Be, Technical is an extremely corrosive acid that comes as a yellowish slightly viscous liquid. It is soluble in water and is a diprotic acid. It has very strong corrosive, dehydrating and oxidizing properties as well as being hygroscopic. It uses range from detergent production, antifreeze, pharmaceuticals and fertilizers to cellophane, lubricants and production of explosives. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 100%
Packaging Glass Bottle
Quantity 2.5 L
Percent Purity ∼93%
Grade Technical
Chemical Name or Material Sulfuric Acid