missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 40% (v/v) Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S7281LT

 View more versions of this product

Catalog No. 18614267


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
40%
1 L
7664-93-9,7732-18-5
Amber Glass Bottle
Ungraded

Chemical Identifiers

7664-93-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.