missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.100N, APHA, Solution, Spectrum™ Chemical

$86.50 - $541.72

Chemical Identifiers

CAS 7664-93-9
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614216
View Documents
Spectrum Chemical Mfg Cor
S615500ML
500 mL Cubitainer
Each for $86.50
Only null left
 
18614300
View Documents
Spectrum Chemical Mfg Cor
S6151LT
1 L Cubitainer
Each for $164.22
Only null left
 
18614301
View Documents
Spectrum Chemical Mfg Cor
S6154LTPL
4 L Cubitainer
Each for $310.50
Only null left
 
18614215
View Documents
Spectrum Chemical Mfg Cor
S61520LT
20 L Cubitainer
Each for $541.72
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7664-93-9

Specifications

7664-93-9,7732-18-5
Cubitainer
APHA/EPA
0.100 N ± 0.001 N
500 mL
Sulfuric Acid
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.