missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 0.05N Standardized Solution

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

$88.79 - $123.58

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45048K2
View Documents
Thermo Scientific Chemicals
045048K2
1 L
Each for $88.79
Only null left
 
AA45048K7
View Documents
Thermo Scientific Chemicals
045048K7
4 L
Each for $123.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O

Specifications

Colourless solution
7664-93-9,7732-18-5
H2O4S
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08
Colorless
1 L
Liquid
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Sulfuric acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-639-5

RTECSNumber : WS5600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only