missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.02N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02004360
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004360 4 L Cubetainer
02004359 1 L Narrow Mouth Polyethylene Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004360 Supplier Avantor Supplier No. 569303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Packaging Cubetainer
Specific Gravity 1L = 1.02kg
Quantity 4 L
Formula Weight 98.08
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Sulfuric Acid, 0.02N Volumetric Solution