missing translation for 'onlineSavingsMsg'
Learn More

Sulfur trioxide-trimethylamine complex, 95%

Catalog No. p-7043337
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0030214 25 g
AAL0030222 100 g
AAL0030236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0030214 Supplier Thermo Scientific Chemicals Supplier No. L0030214
Only null left

CAS: 3162-58-1 | C3H9NO3S | 139.17 g/mol

Sulfur trioxide-trimethylamine complex acts as a reagent for sulfonation and sulfamation reactions. It is used as a reactant for the synthesis of sulfate-conjugated resveratrol metabolites and chitooligosaccharides. It is used in association with dodecyl thioglycopyranoside, which acts as a surfactant for enantiomeric separation. Further, it acts as a nucleophile used for the preparation of alfa-tosyloxy ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfur trioxide-trimethylamine complex acts as a reagent for sulfonation and sulfamation reactions. It is used as a reactant for the synthesis of sulfate-conjugated resveratrol metabolites and chitooligosaccharides. It is used in association with dodecyl thioglycopyranoside, which acts as a surfactant for enantiomeric separation. Further, it acts as a nucleophile used for the preparation of alfa-tosyloxy ketones.

Solubility
Soluble in hot water and ethanol. Slightly soluble in dimethyl sulfoxide and methanol. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 3162-58-1
Molecular Formula C3H9NO3S
Molecular Weight (g/mol) 139.17
MDL Number MFCD00012421
InChI Key DXASQZJWWGZNSF-UHFFFAOYSA-N
Synonym sulfur trioxide trimethylamine complex, sulfur trioxide trimethylamine, sulfur trioxide-trimethylamine complex, sulphur trioxide trimethylamine 1:1, sulfur trioxide-trimethylamine, trimethylamine sulfur trioxide, sulfur trioxide; trimethylamine, 1n1&1&&so3 complex, trimethylamine, compd. with sulfur trioxide 1:1, trimethylamine, compound with sulphur trioxide
PubChem CID 222852
IUPAC Name N,N-dimethylmethanamine;sulfur trioxide
SMILES CN(C)C.O=S(=O)=O

Specifications

Melting Point ∽235°C (decomposition)
Linear Formula (CH3)3N:SO3
Quantity 25 g
UN Number UN3261
Beilstein 3681759
Sensitivity Moisture sensitive
Solubility Information Soluble in hot water and ethanol. Slightly soluble in dimethyl sulfoxide and methanol. Insoluble in water.
Formula Weight 139.17
Percent Purity 95%
Chemical Name or Material Sulfur trioxide-trimethylamine complex
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 221-614-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only