missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, Sublimed, Reagents

Catalog No. p-200021003
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336937 3 kg
01336936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336937 Supplier Labchem Inc Supplier No. C23355003A5
Only null left

Sulfur, Sublimed, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym Sulfur flour, Brimstone
IUPAC Name sulfur
SMILES [S]

Specifications

Boiling Point 444.6°C
Melting Point 106.8°C
Color Yellow
Physical Form Solid
Quantity 3 kg
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.06
Chemical Name or Material Sulfur, Sublimed