missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Oil Standard in #2 Diesel Fuel, 75μg/g, SPEX CertiPrep™

Catalog No. SDFS75Y
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Triple-leached LDPE bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SDFS75Y 100 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDFS75Y Supplier SPEX CertiPrep Group LLC Supplier No. SDFS75Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S75μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 544-40-1 , 68476-34-6
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00009468
InChI Key HTIRHQRTDBPHNZ-UHFFFAOYSA-N
Synonym dibutyl sulfide, butyl sulfide, n-butyl sulfide, butylthiobutane, di-n-butyl sulfide, butane, 1,1'-thiobis, butyl monosulfide, dibutyl thioether, thiononane-5, n-dibutyl sulfide
PubChem CID 11002
IUPAC Name 1-butylsulfanylbutane
SMILES CCCCSCCCC

Specifications

CAS Max % 99.99, 0.01
Physical Form Liquid
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 75μg/g
Packaging Triple-leached LDPE bottle
Chemical Name or Material Sulfur Oil Standard in #2 Diesel Fuel, For Diesel Fuel Analysis
Hazard Category Causes skin irritation, serious eye irritation
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only