missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Oil Standard in #2 Diesel Fuel, 1000μg/g, Set, SPEX CertiPrep™

Catalog No. SDFSSET
Change view
Click to view available options
Quantity:
8 x 125 mL
Packaging:
Triple-leached LDPE bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SDFSSET 8 x 125 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDFSSET Supplier SPEX CertiPrep Group LLC Supplier No. SDFSSET

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Sulfur Oil Standard Set in #2 Diesel Fuel. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Sulfur Oil Standard Set
  • For use with ultra low sulfur standards
  • Includes sulfur standards in #2 Diesel Oil in concentrations of 5, 10, 15, 20, 25, 50 and 100 μg/g
  • Also includes Sulfur Blank

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Sulfur Oil Standards in #2 Diesel Fuel
Name Note For Diesel Fuel Analysis
Physical Form Liquid
Quantity 8 x 125 mL
Packaging Triple-leached LDPE bottle

Product intended for laboratory use only