missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur in Light Mineral Oil standard solution, Specpure™, 1,000μg/g (0.10%)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 mL |
| Grade | Specpure |
| Chemical Name or Material | Sulfur in Light Mineral Oil standard solution, 1,000μg/g (0.10%) |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H350 May cause cancer. |
| Precautionary Statement | GHS P Statement P281-P201-P202-P308+P313-P405-P501a Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. IF exposed or concerned: Get medical advice/attention. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only