Learn More
Minor Rubber Sulfur-Free Nitrometer Tubing
Catalog No. 1417320
Change view
Click to view available options
Color: tan; I.D.: 0.25 in.; O.D.: 0.625 in.; Thickness: 4.8mm
Catalog No.
1417320 Color: tan; I.D.: 0.25 in.; O.D.: 0.625 in.; Thickness: 4.8mm
1 options
Catalog No. 1417320 Supplier Minor Rubber Supplier No. 019083
For connections containing mercury.Made from pure gum rubber. Color, tan.
Order Info
In pack of 50 ft. (15.2m).
Specifications
Diameter (English) Inner 1/4 in.