missing translation for 'onlineSavingsMsg'
Learn More

Sulforhodamine B sodium salt

Catalog No. AAA1476914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1476914 25 g
AAA1476906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1476914 Supplier Thermo Scientific Chemicals Supplier No. A1476914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3520-42-1 | C29H33N2NaO7S2 | 608.70 g/mol

Fluorescent dye which uses laser-induced fluorescence for quantification of cellular proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3520-42-1
Molecular Formula C29H33N2NaO7S2
Molecular Weight (g/mol) 608.70
MDL Number MFCD00010180
InChI Key XJENLUNLXRJLEZ-UHFFFAOYSA-M
Synonym C.I. 45100; Kiton Red S
PubChem CID 131852807
IUPAC Name 2-[3-(diethylamino)-6-diethylazaniumylidenexanthen-9-yl]-5-sulfobenzenesulfonate;sodium
SMILES [Na+].CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C2C=C1C

Specifications

Quantity 25 g
Beilstein 3886008
Formula Weight 580.66
Chemical Name or Material Sulforhodamine B sodium salt
EINECSNumber 222-529-8
RTECSNumber BP6750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only