missing translation for 'onlineSavingsMsg'
Learn More

Sulfonfluorescein 75.0+%, TCI America™

Catalog No. S01321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S01321G 1 g
S01325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S01321G Supplier TCI America Supplier No. S01321G

Chemical Identifiers

CAS 4424-03-7
Molecular Formula C19H12O6S
Molecular Weight (g/mol) 368.36
MDL Number MFCD00070618
InChI Key VJYGFSGAAOVTLC-UHFFFAOYSA-N
PubChem CID 78140
IUPAC Name 1,1-dioxospiro[2,1$l^{6}-benzoxathiole-3,9'-xanthene]-3',6'-diol
SMILES C1=CC=C2C(=C1)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OS2(=O)=O

Specifications

Quantity 1 g
Formula Weight 368.36
Percent Purity ≥75.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfonfluorescein
RTECSNumber WG9930000
TSCA No