missing translation for 'onlineSavingsMsg'
Learn More

Sulfolane, 80% w/w aq. soln.

Catalog No. p-7031431
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32505AP 500 mL
AAH325050F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32505AP Supplier Thermo Scientific Chemicals Supplier No. H32505AP
Only null left

CAS: 126-33-0 | C4H8O2S | 120.17 g/mol

Sulfolane is used in the chemical industry as a solvent for extractive distillation and chemical reactions. It is used to separate aromatic hydrocarbons (benzene, toluene and xylenes), to separate n-propyl alcohol and sec-butyl alcohol. It is also used to purify natural gas streams and for fractionation of fatty acids into saturated and unsaturated components and as a reaction solvent for the preparation of aromatic sulfonic acids, pyridines, isocyanates and pharmaceuticals. It can also be involved in the halogen exchange process and polymerization process. It is used as a plasticizer and curing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfolane is used in the chemical industry as a solvent for extractive distillation and chemical reactions. It is used to separate aromatic hydrocarbons (benzene, toluene and xylenes), to separate n-propyl alcohol and sec-butyl alcohol. It is also used to purify natural gas streams and for fractionation of fatty acids into saturated and unsaturated components and as a reaction solvent for the preparation of aromatic sulfonic acids, pyridines, isocyanates and pharmaceuticals. It can also be involved in the halogen exchange process and polymerization process. It is used as a plasticizer and curing agent.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 126-33-0
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00005484
InChI Key HXJUTPCZVOIRIF-UHFFFAOYSA-N
Synonym sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
PubChem CID 31347
ChEBI CHEBI:74794
IUPAC Name thiolane 1,1-dioxide
SMILES O=S1(=O)CCCC1

Specifications

Melting Point 0°C to 5°C
Density 1.212
Odor Weak, Characteristic
Quantity 500 mL
Beilstein 107765
Merck Index 14,8955
Solubility Information Fully miscible in water.
Formula Weight 120.17
Concentration or Composition (by Analyte or Components) 80% w/w aq. soln.
Chemical Name or Material Sulfolane
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-783-1
RTECSNumber XN0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only