Learn More
Thermo Scientific Chemicals Sulfobromophthalein sodium salt hydrate
CAS: 71-67-0 | C20H8Br4Na2O10S2 | 837.99 g/mol
Supplier: Thermo Scientific Chemicals 04387806
Description
Sulfobromophthalein sodium salt hydrate is a reagent for the quantitative determination of proteins, a dye for liver diagnostics in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sulfobromophthalein sodium salt hydrate | |
| White | |
| MFCD00150017 | |
| Moisture sensitive | |
| bromosulfophthalein, sulfobromophthalein, 3h bsp, 3h-bsp, d00ady | |
| GHAFORRTMVIXHS-UHFFFAOYSA-L | |
| disodium;2-sulfo-4-[4,5,6,7-tetrabromo-1-(4-oxido-3-sulfophenyl)-3-oxo-2-benzofuran-1-yl]phenolate | |
| 102371197 | |
| 837.98 (Anhydrous) |
| 71-67-0 | |
| C20H8Br4Na2O10S2 | |
| 5 g | |
| 14,8954 | |
| Soluble in water. | |
| [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C(=C1)S([O-])(=O)=O | |
| 837.99 | |
| CHEBI:63827 | |
| Powder |
Chemical Identifiers
| 71-67-0 | |
| 837.99 | |
| GHAFORRTMVIXHS-UHFFFAOYSA-L | |
| 102371197 | |
| disodium;2-sulfo-4-[4,5,6,7-tetrabromo-1-(4-oxido-3-sulfophenyl)-3-oxo-2-benzofuran-1-yl]phenolate |
| C20H8Br4Na2O10S2 | |
| MFCD00150017 | |
| bromosulfophthalein, sulfobromophthalein, 3h bsp, 3h-bsp, d00ady | |
| CHEBI:63827 | |
| [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C(=C1)S([O-])(=O)=O |
Safety and Handling
EINECSNumber : 200-761-0
RTECSNumber : SM8395000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only