missing translation for 'onlineSavingsMsg'
Learn More

Sulfobromophthalein sodium hydrate

CAS: 123359-42-2 | C20H8Br4Na2O10S2 | 837.99 g/mol

$762.96 - $762.96

Chemical Identifiers

CAS 123359-42-2
Molecular Formula C20H8Br4Na2O10S2
Molecular Weight (g/mol) 837.99
MDL Number MFCD16556312
InChI Key GHAFORRTMVIXHS-UHFFFAOYSA-L
Synonym disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate
PubChem CID 16211419
IUPAC Name disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate
SMILES [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC173400050
SDS View Documents
Thermo Scientific Chemicals
173400050
5 g Glass bottle
Each for $762.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123359-42-2
837.99
GHAFORRTMVIXHS-UHFFFAOYSA-L
16211419
[Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O
C20H8Br4Na2O10S2
MFCD16556312
disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate
disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate

Specifications

123359-42-2
Authentic
C20H8Br4Na2O10S2
5 g
Solubility in water: soluble
[Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O
837.99
837.76
Crystalline Powder
White
Glass bottle
MFCD16556312
disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate
GHAFORRTMVIXHS-UHFFFAOYSA-L
disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate
16211419
≥95%
Sulfobromophthalein sodium hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protecti

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only