Learn More
Abcam Inc Sulfobetaine 3-16, Zwitterionic detergent, 25G
Catalog No. 502706330
Encompass
MW 391.7 Da, Purity >98%. Zwitterionic detergent. Solubilizes membrane proteins. Retains zwitterionic characteristics over wide pH range.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502706330 Supplier Abcam Inc Supplier No. AB14210325G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 391.7 Da, Purity >98%. Zwitterionic detergent. Solubilizes membrane proteins. Retains zwitterionic characteristics over wide pH range.
The product is subject to the following: Abcam Restricted Use Statement