missing translation for 'onlineSavingsMsg'
Learn More

Sulfo Orange Indicator, 0.04% (w/v), Ricca Chemical

Catalog No. 81134
Change view
Click to view available options
Package: Amber Glass; Size: 120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
81134 Package: Amber Glass; Size: 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 81134 Supplier Ricca Chemical Company Supplier No. 81134

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Sulfo Orange Indicator
CAS 547-57-9, 7732-18-5
CAS Min % 98.0, 0.04
CAS Max % 100.0, 0.04
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Color Orange-Red
Quantity 120 mL