missing translation for 'onlineSavingsMsg'
Learn More

Sulfo Orange Indicator, 0.04% (w/v), Ricca Chemical

Catalog No. 811316
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
811316 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 811316 Supplier Ricca Chemical Company Supplier No. 811316

missing translation for 'validMainframeMsg'

CAS: 547-57-9

Specifications

Chemical Name or Material Sulfo Orange
CAS 547-57-9, 7732-18-5
CAS Min % 98.0, 0.04
CAS Max % 100.0, 0.04
Concentration 0.04% (w/v)
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Color Orange/Red
pH >12
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.04% W/V
Grade Laboratory
Show More Show Less