Learn More
Abcam Inc Sulfo-NHS-LC-Biotin, biotinylating reagent, 50MG
Catalog No. 502705111
Encompass
MW 556.6 Da, Purity >90%. Sulfo-NHS-LC-Biotin, biotinylating reagent. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705111 Supplier Abcam Inc Supplier No. AB14561150MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 556.6 Da, Purity >90%. Sulfo-NHS-LC-Biotin, biotinylating reagent. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement