MW 443.4 Da, Purity >85%. Membrane impermeable, primary amine reactive, short chain (13.5 angstroms) NHS-ester biotinylation reagent. Targets cell surface proteins. Forms irreversible amide bonds. Biotinylates antibodies.
The product is subject to the following: Abcam Restricted Use Statement