missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethoxazole USP , Medisca™

Supplier: Medisca Pharmaceutique Inc 026109
| Quantity | 1 kg |
|---|
Description
Sulfamethoxazole USP is a white to off-white, practically odorless, crystalline powder.
Chemical Identifiers
| 723-46-6 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 723-46-6 | |
| 1 kg | |
| USP |
| Crystalline Powder | |
| White to Off-White | |
| 98.0 to 102.0% |