missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxazole, 98%

Sulfamethoxazole, CAS # 723-46-6, is a sulfonamide antibiotic that inhibits folic acid synthesis in bacteria and prevents cell division and growth. | CAS: 723-46-6 | C10H11N3O3S | 253.276 g/mol

Supplier:  Thermo Scientific Chemicals J6656522

Catalog No. AAJ6656522


Only null left
Explore available promotions

Description

Description

A broad spectrum bacteriostatic sulfonamide antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sulfamethoxazole is an antibiotic composed of an isoxazole with a 4-aminobenzenesulfonamido and methyl-group substitution at positions 5 and 3, respectively
  • Sulfamethoxazole carries out most of its action through the inhibition of dihydropteroate synthase, the enzyme responsible for the synthesis of the precursor of folic acid; inhibition of folic acid synthesis prevents DNA replication and cell division in susceptible bacteria

Application

  • Sulfamethoxazole can elicit an allergic response and may be useful for immune cell studies in laboratory settings
  • Sulfamethoxazole is suitable for laboratory research on infection, environmental pollution, and the physiology of susceptible bacteria
Specifications

Specifications

Sulfamethoxazole
168°C to 172°C
>110°C (230°F)
C10H11N3O3S
100 g
Light sensitive
Soluble in ethanol or acetone. Very slightly soluble in water
CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N
253.276
CHEBI:9332
98%
723-46-6
482°C
Plastic bottle
MFCD00010546
6732984
sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin
JLKIGFTWXXRPMT-UHFFFAOYSA-N
4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
5329
253.28
Powder

Chemical Identifiers

723-46-6
253.276
JLKIGFTWXXRPMT-UHFFFAOYSA-N
5329
4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
C10H11N3O3S
MFCD00010546
sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin
CHEBI:9332
CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 211-963-3

RTECSNumber : WP0700000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

RUO – Research Use Only