missing translation for 'onlineSavingsMsg'
Learn More

Sulfachloropyridazine sodium, 97%, Thermo Scientific™

Catalog No. p-7077711
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458060010 1g
AC458060050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458060010 Supplier Thermo Scientific Chemicals Supplier No. 458060010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23282-55-5
Molecular Formula C10H8ClN4NaO2S
Molecular Weight (g/mol) 306.7
InChI Key GHQJXOBLWKFNAD-UHFFFAOYSA-N
PubChem CID 87077948
IUPAC Name 4-amino-N-(6-chloropyridazin-3-yl)benzenesulfonamide;sodium
SMILES C1=CC(=CC=C1N)S(=O)(=O)NC2=NN=C(C=C2)Cl.[Na]

Specifications

Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 306.7
Percent Purity 97%
Chemical Name or Material Sulfachloropyridazine sodium
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 245-553-

RUO – Research Use Only