missing translation for 'onlineSavingsMsg'
Learn More

Sudan Orange G, Spectrum™ Chemical

Catalog No. p-7166398
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614149 5 g Amber Glass Bottle
18614148 25 g Amber Glass Bottle
18614147 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18614149 Supplier Spectrum Chemical Mfg Cor Supplier No. S16345GM
Only null left

Chemical Identifiers

CAS 2051-85-6

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color Brown to Red
Quantity 5 g
Chemical Name or Material Sudan Orange G

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.