missing translation for 'onlineSavingsMsg'
Learn More

Sudan IV, TCI America™

Supplier:  TCI America O010025G

Catalog No. O010025G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sudan IV
C24H20N4O
Bieblich Scarlet R, Oil Red, Scarlet Red, Scharlach Red, Solvent Red 24
CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C
380.45
380.45
Crystal/Powder
85-83-6
MFCD00003893
KMDLOETUWUPGMB-BXCCFQQFSA-N
(1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
6797604
25 g

Chemical Identifiers

85-83-6
380.45
KMDLOETUWUPGMB-BXCCFQQFSA-N
6797604
CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C
C24H20N4O
MFCD00003893
Bieblich Scarlet R, Oil Red, Scarlet Red, Scharlach Red, Solvent Red 24
(1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3088

RTECSNumber : QL5775000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions