missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan IV

Catalog No. AAA1218114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1218114 25 g
AAA1218122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1218114 Supplier Thermo Scientific Chemicals Supplier No. A1218114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85-83-6 | C24H20N4O | 380.45 g/mol

Useful as a stain for fat in animal tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85-83-6
Molecular Formula C24H20N4O
Molecular Weight (g/mol) 380.45
MDL Number MFCD00003893
InChI Key KMDLOETUWUPGMB-BXCCFQQFSA-N
Synonym C.I. 26105; Scarlet Red
PubChem CID 6797604
IUPAC Name (1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
SMILES CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C

Specifications

Melting Point ∼185°C (decomposition)
Merck Index 14,8393
Quantity 25 g
Formula Weight 380.45
Chemical Name or Material Sudan IV
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 201-635-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only