missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan I, pure

Catalog No. AC147060250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147060250 25 g Glass bottle
AC147061000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC147060250 Supplier Thermo Scientific Chemicals Supplier No. 147060250
Only null left

CAS: 842-07-9 | C16H12N2O | 248.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 842-07-9
Molecular Formula C16H12N2O
Molecular Weight (g/mol) 248.285
MDL Number MFCD00003911
InChI Key ZONYAPYTDIVJGG-VLGSPTGOSA-N
Synonym C.I. 12055, 1-Phenylazo-2-naphthol, Solvent Yellow 14
PubChem CID 5858445
IUPAC Name (1Z)-1-(phenylhydrazinylidene)naphthalen-2-one
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC=CC=C32

Specifications

Melting Point 131.0°C to 133.0°C
Beilstein 16, IV, 228
Infrared Spectrum Authentic
Lambda Max at approximately 418 and 476nm (in ethanol)
Formula Weight 248.28
Grade Pure
Packaging Glass bottle
Specific Extinction 500 min. (at 476nm, in ethanol)
Color Brown or Orange to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Sudan I
Show More Show Less
Hazard Category CANCER SUSPECTED AGENT
Hazard Statement GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-668-2
RTECSNumber QL4900000
TSCA TSCA

RUO – Research Use Only