missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan Black B, TCI America™
Specifications
Specifications
| Melting Point | 124°C |
| Color | Black |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 456.55 |
| Grade | Laboratory |
| Chemical Name or Material | Sudan Black B |
Safety and Handling
| EINECSNumber | (5)-3141 |
| RTECSNumber | SD4431500 |
| TSCA | Yes |