missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan Black B

Histochemical stain for chromosomes | CAS: 4197-25-5 | C29H24N6 | 456.553 g/mol

Supplier:  Thermo Scientific Chemicals J6226814

 View more versions of this product

Catalog No. AAJ6226814


Only null left

Description

Description

Histochemical stain for chromosomesSudan Black B is used as a biological stain for lipids. It is a basic black dye utilized in staining triglycerides and lipids off some lipoproteins. Further, it is used in the fingerprint enhancement as well as for detecting fats. In addition to this, it is used in the study of hematological disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histochemical stain for chromosomesSudan Black B is used as a biological stain for lipids. It is a basic black dye utilized in staining triglycerides and lipids off some lipoproteins. Further, it is used in the fingerprint enhancement as well as for detecting fats. In addition to this, it is used in the study of hematological disorders.

Solubility
Soluble in oils, fats, warm petrolatum, paraffin, phenol, ethanol, acetone, benzene, toluene and hydrocarbon. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Sudan Black B
4197-25-5
MFCD00006919
14,8885
Soluble in oils,fats,warm petrolatum,paraffin,phenol,ethanol,acetone,benzene,toluene and hydrocarbon. Insoluble in water.
CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C
456.553
456.55
25 g
118°C to 126°C
C29H24N6
723248
C.I. 26150; Solvent Black 3
YCUVUDODLRLVIC-UHFFFAOYSA-N
(2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
61336
Black to Brown
Powder

Chemical Identifiers

4197-25-5
456.553
YCUVUDODLRLVIC-UHFFFAOYSA-N
61336
CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C
C29H24N6
MFCD00006919
C.I. 26150; Solvent Black 3
(2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-087-1

RTECSNumber : SD4431500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only