missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan Black B

Catalog No. AAJ6226809
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6226809 10 g
AAJ6226814 25 g
AAJ6226822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6226809 Supplier Thermo Scientific Chemicals Supplier No. J6226809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Histochemical stain for chromosomes | CAS: 4197-25-5 | C29H24N6 | 456.553 g/mol

Histochemical stain for chromosomesSudan Black B is used as a biological stain for lipids. It is a basic black dye utilized in staining triglycerides and lipids off some lipoproteins. Further, it is used in the fingerprint enhancement as well as for detecting fats. In addition to this, it is used in the study of hematological disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histochemical stain for chromosomesSudan Black B is used as a biological stain for lipids. It is a basic black dye utilized in staining triglycerides and lipids off some lipoproteins. Further, it is used in the fingerprint enhancement as well as for detecting fats. In addition to this, it is used in the study of hematological disorders.

Solubility
Soluble in oils, fats, warm petrolatum, paraffin, phenol, ethanol, acetone, benzene, toluene and hydrocarbon. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4197-25-5
Molecular Formula C29H24N6
Molecular Weight (g/mol) 456.553
MDL Number MFCD00006919
InChI Key YCUVUDODLRLVIC-UHFFFAOYSA-N
Synonym C.I. 26150; Solvent Black 3
PubChem CID 61336
IUPAC Name (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
SMILES CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C

Specifications

Melting Point 118°C to 126°C
Beilstein 723248
Merck Index 14,8885
Color Black to Brown
Solubility Information Soluble in oils,fats,warm petrolatum,paraffin,phenol,ethanol,acetone,benzene,toluene and hydrocarbon. Insoluble in water.
Quantity 10 g
Physical Form Powder
Formula Weight 456.55
Chemical Name or Material Sudan Black B
EINECSNumber 224-087-1
RTECSNumber SD4431500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only