missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, Ultrapure Bioreagent, J.T. Baker™
Description
- For Density Gradient Centrifugation and Molecular Biology Applications
Specifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Specific Gravity | 1L = 1.59kg |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
| Formula Weight | 342.3 |
| Percent Purity | ≥99.9% |
| Grade | Ultrapure Bioreagent |
| Chemical Name or Material | Sucrose |