missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sucrose stearate

Catalog No. AAJ6684506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6684506 5 g
AAJ6684514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6684506 Supplier Thermo Scientific Chemicals Supplier No. J6684506

CAS: 25168-73-4 | C30H56O12 | 608.766 g/mol

Sucrose stearate is used as an emulsifier in many beauty applications and cosmetics such as makeup products, cleansing formulas. Emulsifiers aid the blending of water and fatty substances, a common ingredient in many foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose stearate is used as an emulsifier in many beauty applications and cosmetics such as makeup products, cleansing formulas. Emulsifiers aid the blending of water and fatty substances, a common ingredient in many foods.

Solubility
Soluble in water and ethanol at 75°C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 25168-73-4
Molecular Formula C30H56O12
Molecular Weight (g/mol) 608.766
MDL Number MFCD00152822
InChI Key SZYSLWCAWVWFLT-UTGHZIEOSA-N
Synonym sucrose stearate, sucrose, 1-stearate, unii-58rp7ju52k, sucrose stearate ester, alpha-d-glucopyranoside, 1-o-1-oxooctadecyl-beta-d-fructofuranosyl, 1-o-stearoyl-beta-d-fructofuranosyl alpha-d-glucopyranoside, unii-274kw0o50m component, unii-d0951ov2o1 component, unii-l98x941w2b component, 2s,3s,4s,5r-3,4-dihydroxy-5-hydroxymethyl-2-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxolan-2-yl methyl octadecanoate
PubChem CID 9898327
IUPAC Name [(2S,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-2-yl]methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC1(C(C(C(O1)CO)O)O)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in water and ethanol at 75°C.
Formula Weight 608.75
Physical Form Powder
Chemical Name or Material Sucrose stearate
EINECSNumber 246-705-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only