missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sucrose palmitate, 90%

CAS: 26446-38-8 | C28H52O12

$77.51 - $434.26

Chemical Identifiers

CAS 26446-38-8
Molecular Formula C28H52O12
MDL Number MFCD00047545
Synonym beta-D-Fructofuranosyl-alpha-D-glucopyranoside monohexadecanoate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6662718
View Documents
Thermo Scientific Chemicals
J6662718
50 g
Each for $77.51
Only null left
 
AAJ6662730
View Documents
Thermo Scientific Chemicals
J6662730
250 g
Each for $263.03
Only null left
 
AAJ6662736
View Documents
Thermo Scientific Chemicals
J6662736
500 g
Each for $434.26
Only null left
 
Description

Description

Sucrose palmitate is used as food emulsifier. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose palmitate is used as food emulsifier. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in DMSO, DMF, ethanol and water

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications

Chemical Identifiers

26446-38-8
C28H52O12
MFCD00047545
beta-D-Fructofuranosyl-alpha-D-glucopyranoside monohexadecanoate

Specifications

26446-38-8
C28H52O12
50 g
Soluble in DMSO, DMF, ethanol and water
90%
Sucrose palmitate
White
MFCD00047545
beta-D-Fructofuranosyl-alpha-D-glucopyranoside monohexadecanoate
508.72
Powder
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 247-706-7

RTECSNumber : LZ5851350

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only