missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sucrose, ( FCC), MP Biomedicals
Manufacturer: Mp Biomedicals 0290471380
Specifications
Sucrose | |
C12H22O11 | |
100 lb | |
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O | |
342.297 | |
CHEBI:17992 | |
FCC |
57-50-1 | |
amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar | |
CZMRCDWAGMRECN-UGDNZRGBSA-N | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
5988 | |
342.3 |
Chemical Identifiers
57-50-1 | |
342.297 | |
amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar | |
CHEBI:17992 | |
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O |
C12H22O11 | |
CZMRCDWAGMRECN-UGDNZRGBSA-N | |
5988 | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Safety and Handling
Recommended Storage : Room Temperature