missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose Fatty Acid Ester, TCI America™
$451.10 - $451.10
Chemical Identifiers
| MDL Number | MFCD00148531 |
|---|---|
| Synonym | Fatty Acid Sucrose Ester, Saccharose Fatty Acid Ester |
Chemical Identifiers
| MFCD00148531 | |
| Fatty Acid Sucrose Ester, Saccharose Fatty Acid Ester |
Specifications
| MFCD00148531 | |
| Fatty Acid Sucrose Ester, Saccharose Fatty Acid Ester | |
| Sucrose Fatty Acid Ester |
| 500 g | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (8)-0067
TSCA : No