missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, (Crystalline), MP Biomedicals

Catalog No. p-3545931
Change view
Click to view available options
Quantity:
1 lb.
25 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN90297801 1 lb.
ICN90297825 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN90297801 Supplier MP Biomedicals Inc Supplier No. 0290297801
Only null left

This is a slightly less pure sucrose than our 190271. It does not reduce Fehling's solution, and is suitable for most routine applications

  • Its a simple carbohydrate.
  • It is used in density gradients, Dielectrophoretic Field-Flow Fractionation running buffer.
  • It is a main component of media and animal diet.
  • Sucrose is a simple carbohydrate, a disaccharide composed of glucose and fructose.
  • It is hydrolyzed to glucose and fructose by dilute acids and by invertase, a yeast enzyme

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
PubChem CID 5988
ChEBI CHEBI:17992
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Quantity 1 lb.
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Formula Weight 342.3
Physical Form Crystalline
Chemical Name or Material Sucrose
Recommended Storage Room Temperature