missing translation for 'onlineSavingsMsg'
Learn More

Sucrose (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical S53

 View more versions of this product

Catalog No. S5-3



Get Up to $1,800 in Chemicals with Purchase
View all promotions
Only null left

Description

Description

Specifications

Specifications

Sucrose
190°C
6.5 to 7.5
0.03% max.
C12H22O11
3 kg
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
CZMRCDWAGMRECN-PWPRYFECNA-N
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
5988
342.3
Solid
57-50-1
White
0.01% max.
Poly Bottle
MFCD00006626
+66.3 to +66.8° (+ or −)
0.005% max. Insoluble Matter
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
342.30
CHEBI:17992
Certified ACS

Chemical Identifiers

57-50-1
342.30
CZMRCDWAGMRECN-PWPRYFECNA-N
5988
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
C12H22O11
MFCD00006626
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
CHEBI:17992
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:1
Instability:0

SDS Documents

Documents

Promotions

Promotions